5-Bromo-4-methoxy-2-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC910758-1G
PC910758-250MG
APOSPC910758-1GEA
854
EUR
APOSPC910758-1G
APOSPC910758-250MG
5-Bromo-4-methoxy-2-(trifluoromethyl)pyridine 95%
5-Bromo-4-methoxy-2-(trifluoromethyl)pyridine
Formel:
C₇H₅BrF₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD18257659 CAS-Nummer: 1256790-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...