2-Chloro-6-(trifluoromethyl)nicotinaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-6-trifluoromethyl-pyridine-3-carbaldehyde
PC910896-1G
PC910896-5G
APOSPC910896-1GEA
81.2
EUR
APOSPC910896-1G
APOSPC910896-5G
2-Chloro-6-(trifluoromethyl)nicotinaldehyde 95%
2-Chloro-6-(trifluoromethyl)nicotinaldehyde
Formel:
C₇H₃ClF₃NO Molecular Weight: 209,553 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09907845 CAS-Nummer: 944900-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...