Ethyl 2-[(3-fluorophenyl)amino]acetate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl (3-fluorophenyl)glycinate

PC910928-1G PC910928-5G
APOSPC910928-1GEA 53.8 EUR
APOSPC910928-1G APOSPC910928-5G
Ethyl 2-[(3-fluorophenyl)amino]acetate 98%
Ethyl 2-[(3-fluorophenyl)amino]acetate
Formel: C₁₀H₁₂FNO₂
Molecular Weight: 197,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11152956
CAS-Nummer: 2573-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR