7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine 95%

Lieferant: Apollo Scientific
Achtung

PC911133-100MG PC911133-250MG
APOSPC911133-100MGEA 335 EUR
APOSPC911133-100MG APOSPC911133-250MG
7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine 95%
7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine
Formel: C₁₀H₁₂FN
Molecular Weight: 165.207 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06739166
CAS-Nummer: 173998-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR