4-Ethoxy-1-fluoro-2-nitrobenzene 97%

Lieferant: Apollo Scientific

PC911165-1G PC911165-5G
APOSPC911165-1GEA 112 EUR
APOSPC911165-1G APOSPC911165-5G
4-Ethoxy-1-fluoro-2-nitrobenzene 97%
4-Ethoxy-1-fluoro-2-nitrobenzene
Formel: C₈H₈FNO₃
Molecular Weight: 185,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014026
CAS-Nummer: 10298-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR