5-Fluoro-4-methyl-1H-indazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC911241-100MG
PC911241-250MG
APOSPC911241-100MGEA
39.3
EUR
APOSPC911241-100MG
APOSPC911241-250MG
5-Fluoro-4-methyl-1H-indazole 98%
5-Fluoro-4-methyl-1H-indazole
Formel:
C₈H₇FN₂ Molecular Weight: 150,153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007843 CAS-Nummer: 105391-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...