3-Bromo-5-chloro-2-fluorobenzoic acid 95%
Lieferant: Apollo Scientific
PC911333-1G
PC911333-250MG
PC911333-5G
APOSPC911333-1GEA
126
EUR
APOSPC911333-1G
APOSPC911333-250MG
APOSPC911333-5G
3-Bromo-5-chloro-2-fluorobenzoic acid 95%
3-Bromo-5-chloro-2-fluorobenzoic acid
Formel:
C₇H₃BrClFO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD21607478 CAS-Nummer: 1269232-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...