(R)-3-(4-Fluorophenyl)-2-hydroxypropanoic acid 95%

Lieferant: Apollo Scientific

PC911368-1G PC911368-250MG
APOSPC911368-1GEA 647 EUR
APOSPC911368-1G APOSPC911368-250MG
(R)-3-(4-Fluorophenyl)-2-hydroxypropanoic acid 95%
(R)-3-(4-Fluorophenyl)-2-hydroxypropanoic acid
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03839851
CAS-Nummer: 124980-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR