7-Fluoroindoline hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC912334-100MG PC912334-250MG
APOSPC912334-100MGEA 50.9 EUR
APOSPC912334-100MG APOSPC912334-250MG
7-Fluoroindoline hydrochloride 97%
7-Fluoroindoline hydrochloride
Formel: C₈H₈FN·ClH
Molecular Weight: 173,615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27923560
CAS-Nummer: 1820711-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR