8,8-Difluoro-2-azaspiro[4.5]decane hydrochloride

Lieferant: Apollo Scientific

PC912339-1G PC912339-250MG PC912339-500MG
APOSPC912339-1GEA 1550 EUR
APOSPC912339-1G APOSPC912339-250MG APOSPC912339-500MG
8,8-Difluoro-2-azaspiro[4.5]decane hydrochloride
8,8-Difluoro-2-azaspiro[4.5]decane hydrochloride
Formel: C₉H₁₅F₂N·ClH
Molecular Weight: 211,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975525
CAS-Nummer: 1780964-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR