4-Amino-2,3-difluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxy-2,3-difluoroaniline

PC9169-10G PC9169-1G PC9169-5G PC9169-25G
APOSPC9169-10GEA 484 EUR
APOSPC9169-10G APOSPC9169-1G APOSPC9169-5G APOSPC9169-25G
4-Amino-2,3-difluorobenzoic acid
4-Amino-2,3-difluorobenzoic acid
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04114097
CAS-Nummer: 194804-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR