Perfluoroheptanoic anhydride 98%

Lieferant: Apollo Scientific
Gefahr

PC9291-25G PC9291-100G PC9291-5G
APOSPC9291-25GEA 480 EUR
APOSPC9291-25G APOSPC9291-100G APOSPC9291-5G
Perfluoroheptanoic anhydride 98%
Perfluoroheptanoic anhydride
Formel: C₁₄F₂₆O₃
Molecular Weight: 710.106 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00156011
CAS-Nummer: 78225-99-7
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR