4-Iodo-3-(trifluoromethyl)benzoic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Carboxy-2-iodobenzotrifluoride
PC9306-1G
PC9306-5G
PC9306-25G
APOSPC9306-1GEA
21.9
EUR
APOSPC9306-1G
APOSPC9306-5G
APOSPC9306-25G
4-Iodo-3-(trifluoromethyl)benzoic acid 98%
4-Iodo-3-(trifluoromethyl)benzoic acid
Formel:
C₈H₄F₃IO₂ Schmelzpunkt: 194,2…194.8 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09258947 CAS-Nummer: 914636-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...