2,5-Difluorophenylacetyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2,5-Difluorophenyl)acetyl chloride, 2-(2,5-Difluorophenyl)ethanoyl chloride
PC9336-1G
PC9336-5G
APOSPC9336-1GEA
161
EUR
APOSPC9336-1G
APOSPC9336-5G
2,5-Difluorophenylacetyl chloride
2,5-Difluorophenylacetyl chloride
Formel:
C₈H₅ClF₂O Molecular Weight: 190,575 g/mol Siedepunkt: 86…88 °C (11 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD03424719 CAS-Nummer: 157033-23-3 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...