3-[4-(4-Fluorophenyl)piperazin-1-yl]-3-iminopropionitrile 97%

Lieferant: Apollo Scientific
Achtung

PC9363-250MG PC9363-500MG
APOSPC9363-250MGEA 406 EUR
APOSPC9363-250MG APOSPC9363-500MG
3-[4-(4-Fluorophenyl)piperazin-1-yl]-3-iminopropionitrile 97%
3-[4-(4-Fluorophenyl)piperazin-1-yl]-3-iminopropionitrile
Formel: C₁₃H₁₅FN₄
Molecular Weight: 246,283 g/mol
Schmelzpunkt: 157…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664760
CAS-Nummer: 338422-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR