6-(Trifluoromethyl)quinoline-4-thiol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Mercapto-6-(trifluoromethyl)quinoline
PC9444-1G
PC9444-500MG
APOSPC9444-1GEA
473
EUR
APOSPC9444-1G
APOSPC9444-500MG
6-(Trifluoromethyl)quinoline-4-thiol
6-(Trifluoromethyl)quinoline-4-thiol
Formel:
C₁₀H₆F₃NS Molecular Weight: 229,222 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269611 CAS-Nummer: 544716-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...