4-Chloro-3-(trifluoromethyl)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-4'-chloro-3'-(trifluoromethyl)acetophenone, 5-(Bromoacetyl)-2-chlorobenzotrifluoride, 2-Bromo-1-(4-chloro-3-(trifluoromethyl)phenyl)ethanone, 2-Bromo-1-[4-chloro-3-(trifluoromethyl)phenyl]ethan-1-one

PC9516-1G PC9516-5G PC9516-25G
APOSPC9516-1GEA 92.8 EUR
APOSPC9516-1G APOSPC9516-5G APOSPC9516-25G
4-Chloro-3-(trifluoromethyl)phenacyl bromide
4-Chloro-3-(trifluoromethyl)phenacyl bromide
Formel: C₉H₅BrClF₃O
Molecular Weight: 301.488 g/mol
Schmelzpunkt: 36…38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094558
CAS-Nummer: 630404-09-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR