4-(Difluoromethoxy)benzylhydrazine hydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Difluoromethyl 4-(hydrazinomethyl)phenyl ether hydrochloride, 1-(Difluoromethoxy)-4-(hydrazinomethyl)benzene hydrochloride
PC9519-1G
APOSPC9519-1GEA
0
EUR
APOSPC9519-1G
4-(Difluoromethoxy)benzylhydrazine hydrochloride
4-(Difluoromethoxy)benzylhydrazine hydrochloride
Formel:
C₈H₁₀F₂N₂O·ClH Molecular Weight: 224,636 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458142 CAS-Nummer: 887596-65-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...