2,2-Dichloro-3,3,3-trifluoropropan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2-Dichloro-2-(trifluoromethyl)ethan-1-ol, 2,2-Dichloro-3-hydroxy-1,1,1-trifluoropropane
PC9533-10G
PC9533-50G
APOSPC9533-10GEA
72.5
EUR
APOSPC9533-10G
APOSPC9533-50G
2,2-Dichloro-3,3,3-trifluoropropan-1-ol
2,2-Dichloro-3,3,3-trifluoropropan-1-ol
Formel:
C₃H₃Cl₂F₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039302 CAS-Nummer: 20411-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...