2-Amino-5-bromo-3-chlorobenzotrifluoride 98%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2-chloro-6-(trifluoromethyl)aniline
PC9559-1G
PC9559-25G
PC9559-5G
APOSPC9559-1GEA
21.9
EUR
APOSPC9559-1G
APOSPC9559-25G
APOSPC9559-5G
2-Amino-5-bromo-3-chlorobenzotrifluoride 98%
2-Amino-5-bromo-3-chlorobenzotrifluoride
Formel:
C₇H₄BrClF₃N Molecular Weight: 274,466 g/mol Dichte: 1,769 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07369770 CAS-Nummer: 870703-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...