(Z)-1-Bromo-3,3,3-trifluoroprop-1-ene 97%

Lieferant: Apollo Scientific
Achtung

PC9762-1G PC9762-100MG PC9762-250MG
APOSPC9762-1GEA 676 EUR
APOSPC9762-1G APOSPC9762-100MG APOSPC9762-250MG
(Z)-1-Bromo-3,3,3-trifluoroprop-1-ene 97%
(Z)-1-Bromo-3,3,3-trifluoroprop-1-ene
Formel: C₃H₂BrF₃
Molecular Weight: 174,947 g/mol
Siedepunkt: 57…58 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04037239
CAS-Nummer: 149597-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR