Perfluoro-2,5,8,11-tetraoxadodecane 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Perfluorotriglyme, Perfluoro(triethylene glycol dimethyl ether), 1,1,1,3,3,4,4,6,6,7,7,9,9,10,10,12,12,12-Octadecafluoro-2,5,8,11-tetraoxadodecane

PC9775-25G PC9775-5G
APOSPC9775-25GEA 123 EUR
APOSPC9775-25G APOSPC9775-5G
Perfluoro-2,5,8,11-tetraoxadodecane 98%
Perfluoro-2,5,8,11-tetraoxadodecane
Formel: C₈F₁₈O₄
Molecular Weight: 502,054 g/mol
Siedepunkt: 106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458174
CAS-Nummer: 64028-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR