3-Fluoropiperidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC9782-1G
PC9782-2.5G
PC9782-5G
APOSPC9782-1GEA
125
EUR
APOSPC9782-1G
APOSPC9782-2.5G
APOSPC9782-5G
3-Fluoropiperidine hydrochloride
3-Fluoropiperidine hydrochloride
Formel:
C₅H₁₀FN·HCl Molecular Weight: 139.60 g/mol Schmelzpunkt: 204 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD03452799 CAS-Nummer: 116574-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...