1-(2-Chloro-6-fluorobenzyl)homopiperazine dihydrochloride
Lieferant: Apollo Scientific
PC9840-2G
PC9840-10G
APOSPC9840-2GEA
183
EUR
APOSPC9840-2G
APOSPC9840-10G
1-(2-Chloro-6-fluorobenzyl)homopiperazine dihydrochloride
1-(2-Chloro-6-fluorobenzyl)homopiperazine dihydrochloride
Formel:
C₁₂H₁₆ClFN₂·2ClH Molecular Weight: 315,642 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973341 CAS-Nummer: 1049733-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...