Ethyl 3-chloro-2-fluoro-4-methoxybenzoate 97%

Lieferant: Apollo Scientific

PC99022-1G PC99022-500MG
APOSPC99022-1GEA 886 EUR
APOSPC99022-1G APOSPC99022-500MG
Ethyl 3-chloro-2-fluoro-4-methoxybenzoate 97%
Ethyl 3-chloro-2-fluoro-4-methoxybenzoate
Formel: C₁₀H₁₀ClFO₃
Molecular Weight: 232,636 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32707291
CAS-Nummer: 2112478-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR