3-[2-(Trifluoromethyl)phenyl]-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
PC9986-1G
PC9986-250MG
APOSPC9986-1GEA
219
EUR
APOSPC9986-1G
APOSPC9986-250MG
3-[2-(Trifluoromethyl)phenyl]-1H-pyrazole
3-[2-(Trifluoromethyl)phenyl]-1H-pyrazole
Formel:
C₁₀H₇F₃N₂ Molecular Weight: 212,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569437 CAS-Nummer: 149739-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...