3-[4-(Trifluoromethoxy)phenyl]-1H-pyrazole 97%

Lieferant: Apollo Scientific
Achtung

PC9992-250MG PC9992-1G
APOSPC9992-250MGEA 65.4 EUR
APOSPC9992-250MG APOSPC9992-1G
3-[4-(Trifluoromethoxy)phenyl]-1H-pyrazole 97%
3-[4-(Trifluoromethoxy)phenyl]-1H-pyrazole
Formel: C₁₀H₇F₃N₂O
Molecular Weight: 228,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02091526
CAS-Nummer: 474707-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR