CAPS (3-(Cyclohexylamino)propansulfonsäure) ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Cyclohexylaminopropanesulphonic acid
BI5848-100G
BI5848-1KG
BI5848-250G
APOSBI5848-100GEA
30.6
EUR
APOSBI5848-100G
APOSBI5848-1KG
APOSBI5848-250G
CAPS (3-(Cyclohexylamino)propansulfonsäure) ≥98%
CAPS (3-(Cyclohexylamino)propansulfonsäure)
- Loss on drying <5.0%
- pH (1% aqueous) = 4.9-6.0 at 25°C
- Biological buffer - useful range 9.7-11.1
Formel:
C₆H₁₁NH(CH₂)₃SO₃H Molecular Weight: 221,32 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003837 CAS-Nummer: 1135-40-6 EINECS: 214-492-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...