Bis-TRIS hydrochloride ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol hydrochloride, 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)aminotris(hydroxymethyl)methane hydrochloride
BI5923-100G
BI5923-250G
BI5923-500G
APOSBI5923-100GEA
109
EUR
APOSBI5923-100G
APOSBI5923-250G
APOSBI5923-500G
Bis-TRIS hydrochloride ≥98%
Bis-TRIS hydrochloride
- Buffer
Formel:
C₈H₂₀NO₅Cl Molecular Weight: 245,7 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00171252 CAS-Nummer: 124763-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...