HEPES hemisodium salt ≥96%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-2-Hydroxyethylpiperazine-N'-2-ethanesulphonic acid hemisodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid hemisodium salt

BI5940-100G BI5940-25G BI5940-500G
APOSBI5940-100GEA 71.2 EUR
APOSBI5940-100G APOSBI5940-25G APOSBI5940-500G
HEPES hemisodium salt ≥96%
HEPES hemisodium salt

Biological buffer.


  • Assay (by titration) 98+%
  • Water (By Karl Fischer) <3.0%
  • pH (1%) = 7.4 +/- 0.2

Formel: C₈H₁₇.₅N₂O₄S·½Na
Molecular Weight: 249.30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036463
CAS-Nummer: 103404-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR