1,4-Phenylendimethanthiol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: alpha,alpha'-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene, 1,4-BDMT

BI7566-250MG BI7566-1G BI7566-5G
APOSBI7566-250MGEA 21.9 EUR
APOSBI7566-250MG APOSBI7566-1G APOSBI7566-5G
1,4-Phenylendimethanthiol 98%
1,4-Benzenedimethanethiol

Bitte beachten Store under Argon.

Formel: C₈H₁₀S₂
Molecular Weight: 170.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004872
CAS-Nummer: 105-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR