4-(3-Indolyl)butansäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Indol-3-buttersäure, 4-(Indol-3-yl)buttersäure
BI9764-25G
BI9764-5G
APOSBI9764-25GEA
65.4
EUR
APOSBI9764-25G
APOSBI9764-5G
4-(3-Indolyl)butansäure
4-(3-Indolyl)butansäure
- Assay (by titration) 99+%
- Water (by Karl Fischer) < 1.0%
- Plant tissue culture tested
Formel:
C₁₂H₁₃NO₂ Molecular Weight: 203.24 g/mol Schmelzpunkt: 122…125 °C Dichte: 1.27 Lagertemperatur: Kühlschrank |
MDL:
MFCD00005664 CAS-Nummer: 133-32-4 EINECS: 205-101-5 UN: 2811 ADR: 6.1 III Merck Index: 13,04987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...