Amoxicillin trihydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-[2-Amino-2-(4-hydroxyphenyl)-acetyl]amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxicillin Ttrihydrate, (2S,5R,6S)-6-((S)-2-amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

BIA0101-10G BIA0101-1G BIA0101-25G
APOSBIA0101-10GEA 37.7 EUR
APOSBIA0101-10G APOSBIA0101-1G APOSBIA0101-25G
Amoxicillin trihydrate
Amoxicillin trihydrate

Inhibitor of bacterial cell wall synthesis.sensitive to b-lactamase.


  • Cell culture tested

Formel: C₁₆H₁₉N₃O₅S
Molecular Weight: 365.40 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00072029
CAS-Nummer: 61336-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR