3-Amino-9-ethylcarbazol (AEC) ≥95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 9-Ethylcarbazol-3-ylamin, AEC

BIA0303-100G BIA0303-25G BIA0303-5G
APOSBIA0303-100GEA 284 EUR
APOSBIA0303-100G APOSBIA0303-25G APOSBIA0303-5G
3-Amino-9-ethylcarbazol (AEC) ≥95%
3-Amino-9-ethyl carbazole (AEC)

Substrate for peroxidase.

Formel: C₁₄H₁₄N₂
Molecular Weight: 210,28 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004964
CAS-Nummer: 132-32-1
EINECS: 205-057-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR