DL-Alanin

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-Ala-OH, (±)-2-Aminopropansäure, (±)-2-Aminopropionic acid, (±)-Alanin, α-Alanin, (±)-2-Aminopropionsäure, DL-α-Alanin

BIA0868-100G BIA0868-500G
APOSBIA0868-100GEA 21.9 EUR
APOSBIA0868-100G APOSBIA0868-500G
DL-Alanin
DL-Alanin

Pyrogen free.

Formel: C₃H₇NO₂
Molecular Weight: 89.09 g/mol
Siedepunkt: 189
Schmelzpunkt: 289 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064408
CAS-Nummer: 302-72-7
EINECS: 206-126-4
Merck Index: 13,00203

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR