N-Succinimidyl Acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate
BIA1015-10G
BIA1015-25G
BIA1015-100G
APOSBIA1015-10GEA
72.5
EUR
APOSBIA1015-10G
APOSBIA1015-25G
APOSBIA1015-100G
N-Succinimidyl Acetate
N-Succinimidyl Acetate
Formel:
C₆H₇NO₄ Molecular Weight: 157,13 g/mol Schmelzpunkt: 134 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00037916 CAS-Nummer: 14464-29-0 EINECS: 238-454-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...