Dansylchlorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Dimethylamino)naphthalinsulfonylchlorid, 5-(Dimethylamino)naphthalinsulfonsäurechlorid, 5-(Dimethylamino)naphthalin-1-sulfonylchlorid

BIA110-1G BIA110-25G BIA110-5G
APOSBIA110-1GEA 21.9 EUR
APOSBIA110-1G APOSBIA110-25G APOSBIA110-5G
Dansylchlorid
Dansylchlorid

Fluorometric detection at picomole levels.

Formel: C₁₂H₁₂ClNO₂S
Molecular Weight: 269,75 g/mol
Schmelzpunkt: 69…75 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00003985
CAS-Nummer: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR