Acridinorange (Zinkchlorid-Doppelsalz)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride
BIA1145-100G
BIA1145-25G
APOSBIA1145-100GEA
79.9
EUR
APOSBIA1145-100G
APOSBIA1145-25G
Acridinorange (Zinkchlorid-Doppelsalz)
Acridinorange (Zinkchlorid-Doppelsalz)
A fluorescent stain for nuclei acids in agarose andpolyacrylamide gels.
Formel:
C₁₇H₂₀ClN₃·HCl·½ZnCl₂ Molecular Weight: 369,96 g/mol Schmelzpunkt: 165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081043 CAS-Nummer: 10127-02-3 UN: 3143 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...