N-Acetyl-D-mannosamine, Monohydrat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cyclic N-Acetyl-D-mannosamine, N-((2R,3S,4R,5S,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide
BIA1221-10G
BIA1221-25G
APOSBIA1221-10GEA
137
EUR
APOSBIA1221-10G
APOSBIA1221-25G
N-Acetyl-D-mannosamine, Monohydrat
N-Acetyl-D-mannosamine
Formel:
C₈H₁₅NO₆ Molecular Weight: 221,21 g/mol (anhy) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00149493 CAS-Nummer: 7772-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...