N-Acetyl-β-D-glucosamin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
BIA1710-25G
BIA1710-5G
APOSBIA1710-25GEA
132
EUR
APOSBIA1710-25G
APOSBIA1710-5G
N-Acetyl-β-D-glucosamin
N-Acetyl-β-D-glucosamin
From synthetic source.
Formel:
C₈H₁₅NO₆ Molecular Weight: 221,21 g/mol Schmelzpunkt: 211…213 °C (Zersetzung) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00136044 CAS-Nummer: 7512-17-6 EINECS: 231-368-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...