N(α)-Acetyl-L-Tryptophan 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Acetyl-L-tryptophan, Ac-Trp-OH
BIA1792-100G
APOSBIA1792-100GEA
59.6
EUR
APOSBIA1792-100G
N(α)-Acetyl-L-Tryptophan 98%
N(α)-Acetyl-L-tryptophan
Formel:
C₁₃H₁₄N₂O₃ Molecular Weight: 246,3 g/mol Schmelzpunkt: 186 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00065976 CAS-Nummer: 1218-34-4 EINECS: 214-935-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...