Ribit

Lieferant: Apollo Scientific

Synonyme: Ribitol, 1,2,3,4,5-Pentanpentol, Adonitrol, (2R,3s,4S)-1,2,3,4,5-Pentanpentol, Ribit, Adonit, Pentitol

BIA3219-250G BIA3219-50G
APOSBIA3219-250GEA 183 EUR
APOSBIA3219-250G APOSBIA3219-50G
Ribit
Adonitol
Formel: HOH₂C(CH(OH))₃CH₂OH
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064291
CAS-Nummer: 488-81-3
Merck Index: 13,00168

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR