6-Aminoindoline dihydrochloride

Lieferant: Apollo Scientific
Achtung

BIA4008-1G
APOSBIA4008-1GEA 65.4 EUR
APOSBIA4008-1G
6-Aminoindoline dihydrochloride
6-Aminoindoline dihydrochloride
Formel: C₈H₁₀N₂·2HCl
Molecular Weight: 207,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035181
CAS-Nummer: 28228-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR