Acriflavin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base
BIA4303-100G
BIA4303-25G
APOSBIA4303-100GEA
98.6
EUR
APOSBIA4303-100G
APOSBIA4303-25G
Acriflavin 98%
Acriflavin
A mixture of 3,6-diamino-10-methylacridiniumchloride and 3,6-diaminoacridine.
Formel:
C₂₇H₂₅ClN₆ Molecular Weight: 468.9806 g/mol Schmelzpunkt: 179…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064307 CAS-Nummer: 8048-52-0 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...