Aflatoxicol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Aflatoxin R0
BIA4321-1MG
BIA4321-100UG
APOSBIA4321-1MGEA
0
EUR
APOSBIA4321-1MG
APOSBIA4321-100UG
Aflatoxicol
Aflatoxicol
Formel:
C₁₇H₁₄O₆ Molecular Weight: 314.3 g/mol Schmelzpunkt: 268…269 °C Lagertemperatur: Gefrierschrank |
CAS-Nummer:
29611-03-8 EINECS: 249-727-7 UN: 3462 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...