Zebularine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Deoxyuridine, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

BIA8416-100MG BIA8416-50MG
APOSBIA8416-100MGEA 161 EUR
APOSBIA8416-100MG APOSBIA8416-50MG
Zebularine
Zebularine
Formel: C₉H₁₂N₂O₅
Molecular Weight: 228.20 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04973699
CAS-Nummer: 3690-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR