4-Nitrophenyl-β-D-cellobioside

Lieferant: Apollo Scientific
Achtung

Synonyme: Glc beta(1-4)Glc-β-pNP

BIB1431-100MG BIB1431-250MG BIB1431-500MG
APOSBIB1431-100MGEA 72.5 EUR
APOSBIB1431-100MG APOSBIB1431-250MG APOSBIB1431-500MG
4-Nitrophenyl-β-D-cellobioside
4-Nitrophenyl-beta-D-cellobioside
Formel: C₁₈H₂₅NO₁₃
Molecular Weight: 463,39 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00069845
CAS-Nummer: 3482-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR