(4-Nitrophenyl)-α-D-glucopyranosid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside

BIB1434-1G BIB1434-5G BIB1434-25G
APOSBIB1434-1GEA 43.5 EUR
APOSBIB1434-1G APOSBIB1434-5G APOSBIB1434-25G
(4-Nitrophenyl)-α-D-glucopyranosid
(4-Nitrophenyl)-α-D-glucopyranosid
Formel: C₁₂H₁₅NO₈
Molecular Weight: 301,25 g/mol
Schmelzpunkt: 209…213 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00064088
CAS-Nummer: 3767-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR