4-Nitrophenyl-β-D-xylopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: PNPX, 4-Nitrophenyl β-D-xylopyranoside

BIB1436-1G BIB1436-250MG
APOSBIB1436-1GEA 116 EUR
APOSBIB1436-1G APOSBIB1436-250MG
4-Nitrophenyl-β-D-xylopyranoside
4-Nitrophenyl-beta-D-xylopyranoside
Formel: C₁₁H₁₃NO₇
Molecular Weight: 271,23 g/mol
Schmelzpunkt: 158…160 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00047519
CAS-Nummer: 2001-96-9
EINECS: 217-897-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR