Pyr-AMC

Lieferant: Apollo Scientific

Synonyme: L-Pyroglutamic acid 7-amido-4-methylcoumarin

BIB1445-100MG BIB1445-250MG
APOSBIB1445-100MGEA 232 EUR
APOSBIB1445-100MG APOSBIB1445-250MG
Pyr-AMC
Pyr-AMC
Formel: C₁₅H₁₄N₂O₄
Molecular Weight: 286.29 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00080240
CAS-Nummer: 66642-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR